Learn More
Thermo Scientific™ Remel™ BHI Agar w/Sheep Blood, Penicillin, Streptomycin
Thermo Scientific™ Remel BHI agar with Sheep Blood, Penicillin, Streptomycin is for selective isolation of fastidious fungi and yeast.
$131.04 - $323.90
Specifications
Description BHI Agar w/Sheep Blood, Penicillin, Streptomycin
View More Specs
Products 2
Catalog Number Mfr. No. Format Price Quantity & Availability  
Catalog Number Mfr. No. Format Price Quantity & Availability  
R01152
Accessories View Documents
Thermo Scientific™
R01152
Monoplate Deep Pour Pack of 10 for $131.04
Add to cart
R08092
View Documents
Thermo Scientific™
R08092
20mm x 113mm Tube Pack of 20 for $323.90
Add to cart
Description
Description
Thermo Scientific™ Remel BHI agar with Sheep Blood, Penicillin, Streptomycin is for selective isolation of fastidious fungi and yeast.
  • Penicillin inhibits gram-positive bacteria
  • Streptomycin inhibits some gram-positive and gram-negative bacteria
Specifications
Specifications
BHI Agar w/Sheep Blood, Penicillin, Streptomycin