Learn More
Thermo Scientific™ Remel™ BHI Agar w/Sheep Blood, Penicillin, Streptomycin
Thermo Scientific™ Remel BHI agar with Sheep Blood, Penicillin, Streptomycin is for selective isolation of fastidious fungi and yeast.
Manufacturer:  Thermo Scientific™ R01152
 View more versions of this product
Catalog No. R01152
Add to cart
Description
Description
Thermo Scientific™ Remel BHI agar with Sheep Blood, Penicillin, Streptomycin is for selective isolation of fastidious fungi and yeast.
  • Penicillin inhibits gram-positive bacteria
  • Streptomycin inhibits some gram-positive and gram-negative bacteria
Specifications
Specifications
BHI Agar w/Sheep Blood, Penicillin, Streptomycin
Agar
Monoplate Deep Pour
10/Pk.
SDS
Documents
Accessories