Learn More
Thermo Scientific™ Blood Heart Infusion (BHI) Agar with Sheep Blood, Penicilllin and Streptomycin
Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with this nutritious medium.
Supplier: Thermo Scientific™ R08092
Description
Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with the nutritious base medium Thermo Scientific™ Remel™ Blood Heart Infusion (BHI) Agar with Sheep Blood, Penicilllin and Streptomycin 1. Isolate fastidious fungi and grow Actinomyces2 using BHI Agar with 5-10% sheep blood. Pathogenic fungi and Nocardia asteroides can be isolated using optimal concentrations of chloramphenicol3. Cycloheximide inhibits certain Aspergillus spp. and zygomycetes
- Penicillin inhibits gram-positive bacteria
- Streptomycin inhibits some gram-positive and gram-negative bacteria
Specifications
BHI Agar w/Sheep Blood, Penicillin, Streptomycin | |
20mm x 113mm Tube | |
Agar | |
20/Pk. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.