Learn More
Thermo Scientific™ BHI Agar with Sheep Blood
Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with this nutritious medium.
Supplier: Thermo Scientific™ R08064
Description
Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with nutritious Thermo Scientific™ Remel™ BHI Agar with Sheep Blood. Due to the addition of Sheep blood, the BHI Agar can be used for the isolation of dimorphic fungi which may grow poorly on non-enriched media.
Use BHI Agar with Sheep Blood for the cultivation of a wide variety of micro-organisms including pathogenic fungi.
- Highly Nutritious: medium recommended for the enrichment of fastidious organisms.
- Ready-to-use: convenience of prepared media.
BHI Agar w/Sheep Blood contains brain heart infusion, peptone, and dextrose which provide nitrogen, carbon, sulfur, vitamins and a carbohydrate source. Sheep blood provides essential growth factors for fastidious fungi. Yeast extracts are incorporated in the medium to serve as a growth stimulant.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
BHI Agar w/Sheep Blood | |
20mm x 113mm Tube | |
Agar | |
100/Pk. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.