Learn More
Thermo Scientific™ Remel™ Biplates, HE through V Agar Available on GSA/VA Contract for Federal Government customers only.
Perform screening and isolation in the same 12 x 85mm dish, divided into two sections to hold either the same media on both sides or two media, one on each side.
$165.29 - $219.45
Specifications
Format Biplate
Packaging 10/Pk.
View More Specs
Products 6
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
R02086
View Documents
Thermo Scientific™
R02086
HE Agar; XLD Agar
Pack of 10 for $184.19
Add to cart
R02095 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R02095
MacConkey Agar//MacConkey Agar w/Sorbitol
Pack of 10 for $203.81
Add to cart
R02096 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R02096
MacConkey Agar//PEA Sheep Blood Agar
Pack of 10 for $166.11
Add to cart
R02112 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R02112
Middlebrook 7H11 Agar//Mitchison 7H11 Selective
Pack of 10 for $165.29
Add to cart
R02148 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R02148
V Agar; Starch Agar w/Brom Cresol Purple; Biplate; 10/Pk.
Pack of 10 for $181.34
Add to cart
R02140 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R02140
QC-Slide™ AFB Stain Control; 10/Pk.
Pack of 10 for $219.45
Add to cart
Description
Description
Perform screening and isolation in the same 12 x 85mm dish using Thermo Scientific™ Remel Biplates, divided into two sections to hold either the same media on both sides or two media, one on each side.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Biplate
10/Pk.