Learn More
Thermo Scientific™ V Agar/V Agar Selective with Starch Agar and Brom Cresol Purple Biplate
Isolate, presumptively identify, and differentiate Gardnerella vaginalis from other organisms using this Biplate.
Supplier: Thermo Scientific™ R02096
Description
Isolate, presumptively identify, and differentiate Gardnerella vaginalis from other organisms using Thermo Scientific™ Remel™ V Agar and V Agar Selective with Starch Agar and Brom cresol Purple Biplate. Greenwood et al. introduced “V“ (Vaginalis) Agar, a Columbia agar base which contained 5% whole human blood1 and evaluated 78 strains of G. vaginalis for various characteristics including ability to produce beta-hemolysis on human blood agar and sheep blood agar2. More than 90% of strains tested were beta-hemolytic on human blood agar, none were beta-hemolytic on sheep blood agar.
Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.
- Ready-to-use <
Specifications
MacConkey Agar//PEA Sheep Blood Agar | |
Biplate | |
Agar | |
10/Pk. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.