Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Brilliant Green Bile Broth 2%

Thermo Scientific™ Remel Brilliant Green Bile Broth 2% is for the detection of coliform microorganisms in water, wastewater, foods, and dairy products.

$335.00 - $1367.00

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, EPA, IDF, SMWW.
Description Brilliant Green Bile Broth 2%
Format 16mm x 125mm Durham Tube
View More Specs

Products 2
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
R07020
View Documents
Thermo Scientific™
R07020
100/Pk.
Case of 100 for $1,367.00
Add to cart
 
R07022
View Documents
Thermo Scientific™
R07022
20/Pk.
Pack of 20 for $335.00
Add to cart
 
Description

Description

Thermo Scientific™ Remel Brilliant Green Bile Broth 2% is for the detection of coliform microorganisms in water, wastewater, foods, and dairy products.

Fermentation is detected by gas in Durham tube.
Specifications

Specifications

Industrial Reference: AOAC, BAM, EPA, IDF, SMWW.
Brilliant Green Bile Broth 2%
16mm x 125mm Durham Tube
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit