Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ Brilliant Green Bile Broth 2%
Thermo Scientific™ Remel Brilliant Green Bile Broth 2% is for the detection of coliform microorganisms in water, wastewater, foods, and dairy products.
Supplier: Thermo Scientific™ R07020
Description
Thermo Scientific™ Remel Brilliant Green Bile Broth 2% is for the detection of coliform microorganisms in water, wastewater, foods, and dairy products.
Fermentation is detected by gas in Durham tube.Specifications
Industrial Reference: AOAC, BAM, EPA, IDF, SMWW. | |
16mm x 125mm Durham Tube | |
100 x 10 mL |
Brilliant Green Bile Broth 2% | |
Microbiology Media |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction