Learn More
Thermo Scientific™ Brucella Agar with 5% Sheep Blood, Hemin and Vitamin K Available on GSA/VA Contract for Federal Government customers only.
Isolate and cultivate anaerobic bacteria including Brucella, from clinical and non-clinical specimens with this ready-to-use formulation that conforms to APHA.
$184.91 - $750.23
Specifications
Description Brucella Agar w/5% Sheep Blood, Hemin, Vitamin K
View More Specs
Products 2
Catalog Number Mfr. No. Format Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Format Packaging Price Quantity & Availability  
R01255 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R01255
12mm x 85mm Monoplate 100/Pk. Case of 100 for $750.23
Add to cart
R04011 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R04011
15mm x 150mm Plate 10/Pk. Pack of 10 for $184.91
Add to cart
Description
Description
Isolate and cultivate anaerobic bacteria including Brucella, from clinical and non-clinical specimens with Thermo Scientific™ Remel™ Brucella Agar prepared plates with 5% Sheep Blood, Hemin and Vitamin K. Brucella Agar with 5% Sheep Blood, Hemin and Vitamin K is prepared according to the formulation of the American Health Association (APHA)1. Finegold reported Brucella agar with 5% sheep blood is preferable to heart infusion agar for the cultivation of anaerobic bacteria2. The addition of Vitamin K, hemin and sheep blood in the medium makes it appropriate for the cultivation of many anaerobes.
Brucellosis is a highly contagious zoonosis caused by ingestion of unpasteurized milk or undercooked meat from infected animals, or close contact with their secretions.
Brucella Agar is developed for the cultivation of Brucella species from diagnostic specimens such as blood, and from foods and potentially contaminated material.
  • Prepared according to the formulation of the American Health Association (APHA)1.
  • Preferable to Heart Infusion Blood Agar Base for the cultivation of anaerobic bacteria2.
  • Ready-to-use convenience of prepared media.
Brucella Agar with 5% Sheep Blood, Hemin and Vitamin K is a rich nutrient base that supports the growth of fastidious organisms.  The media contains dextrose which provides an energy source; peptones to provide nitrogenous compounds, and yeast extract to supply B vitamins. Growth factors required by some anaerobic bacteria are provided for in the added sheep blood, which also allows the demonstration of hemolytic reactions. Hemin and vitamin K are incorporated into the medium to enhance the growth of gram-positive spore-formers and Bacteroides species.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Brucella Agar w/5% Sheep Blood, Hemin, Vitamin K