Learn More
Thermo Scientific™ Remel™ Calibrated Inoculating Loops
Thermo Scientific™ Remel Calibrated Inoculating Loops come in a variety of materials: platinum, twisted nichrome and plastic.Some with color coded with insulated handle.
Manufacturer:  Thermo Scientific™ R501511
 View more versions of this product
Catalog No. R501511
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Thermo Scientific™ Remel Calibrated Inoculating Loops are available in platinum, twisted nichrome and and plastic. Some are color coded with insulated handle.
  • Twisted nichrome or plastic
  • With or without insulated handle
  • Color-coded
Specifications
Specifications
Plastic
Yellow
32 Per Pack
10µL