Learn More
Thermo Scientific™ Campy Blood Agar Blaser with 5% Sheep Blood and Antibiotics Available on GSA/VA Contract for Federal Government customers only.
Carry out primary and selective isolation of Campylobacter species from fecal specimens with this selective, ready-to-use medium.
$65.42 - $630.53
Specifications
Description Campy Blood Agar Blaser
View More Specs
Products 3
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
R01280 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R01280
10/Pk.
Pack of 10 for $65.42
Add to cart
R01281 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R01281
15/Pk.
Pack of 15 for $118.55
Add to cart
R01284 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R01284
100/Pk.
Case of 100 for $630.53
Add to cart
Description
Description
Carry out primary and selective isolation of Campylobacter species from fecal specimens with the Thermo Scientific™ Remel™ Campy Blood Agar Blaser with 5% Sheep Blood and Antibiotics. Campy Blood Agar Blaser with 5% Sheep Blood and Antibiotics is a modification of Brucella Agar Base with Sheep blood and Antibiotics by Blaser. The additional antibiotic cephalothin was added to improve the inhibition of commensal microbial flora in fecal specimens1.
Campylobacter are Gram-negative, spirally shaped microaerophilic organisms which may be present in raw milk, untreated water, improperly handled food and undercooked meats, poultry and shellfish. Human consumption of these organisms can result in a range of clinical illnesses from transient asymptomatic colonization to severe dysentery. The symptoms of Campylobacter enteritis include diarrhea, stomach pain, nausea, fever, and headache and muscle pain.
Use Campy Blood Agar with 5% Sheep Blood, Antibiotics for primary and selective isolation of Campylobacter from fecal specimens.
  • Selective: the incorporation of cephalothin, amphotericin B, trimethoprim, vancomycin, and polymyxin B in the medium makes the medium inhibitory to Enterobacteriaceae, staphylococci, and yeast.
  • Ready to use: convenience of prepared media.
Casein and meat peptones in the medium provide nitrogen, carbon, and sulfur which are essential for the recovery of Campylobacter sp. Sodium is a source of essential electrolytes and maintains osmotic equilibrium. Dextrose is an energy source and yeast extract supplies B vitamins. Sheep blood provides hemin and other necessary growth factors.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Campy Blood Agar Blaser