Learn More
Thermo Scientific™ Remel™ Campy Thioglycollate Medium (w/.16% Agar) Available on GSA/VA Contract for Federal Government customers only.
Used as a holding medium for samples prior to inoculation of media for recovery of Campylobacter spp.
$219.98 - $1084.23
Specifications
Description Campy Thioglycollate Medium
Format 15mm x 103mm Tube
View More Specs
Products 2
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
R060370 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R060370
100/Pk.
Case of 100 for $1,084.23
Add to cart
R060372 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R060372
20/Pk.
Pack of 20 for $219.98
Add to cart
Description
Description
Thermo Scientific™ Remel Campy Thioglycollate Medium (w/.16% agar, trimethoprim, vancomycin, polymyxin B, cephalothin, amphotericin B) is used as a holding medium for samples prior to inoculation of media for recovery of Campylobacter spp.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Campy Thioglycollate Medium
15mm x 103mm Tube