Learn More
Thermo Scientific™ Remel™ Color Slide™ II Mononucleosis Test Available on GSA/VA Contract for Federal Government customers only.
Rapid slide agglutination test using horse erythrocytes for detection of heterophile antibodies of infectious mononucleosis.
Manufacturer:  Thermo Scientific™ R2468944
 View more versions of this product
Catalog No. 22-286555
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Thermo Scientific™ Remel Color Slide and II Mononucleosis Test is a rapid slide agglutination test using horse erythrocytes for detection of heterophile antibodies of infectious mononucleosis.
  • Dual color indicator
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Color Slide™ II Mononucleosis Test
50 Tests/kit