Learn More
Thermo Scientific™ Remel™ Colorimeter Standard Set, Vitek™
Description
Thermo Scientific™ Remel Colorimeter Standard Kit contains latex equivalence turbidity standards for use with Vitek colorimeter.
Specifications
Specifications
| For Use With (Equipment) | Vitek |
| Description | Colorimeter Standard Set |
| Includes | one each of 0.5, 1.0, 2.0, and 3.0 standards |