Learn More
Thermo Scientific™ Remel™ CTA w/1% Xylose (Cystine Trypticase Agar)
Use for determination of fermentation reactions by fastidious microorganisms, such as Neisseria, pneumococci, streptococci and non-spore forming anaerobes.
Supplier: Thermo Scientific™ R060668
This item is not returnable.
View return policy
Description
Thermo Scientific™ Remel CTA with 1% Xylose (Cystine Trypticase Agar) is used for determination of fermentation reactions by fastidious microorganisms, such as Neisseria, pneumococci, streptococci and non-spore forming anaerobes.
Specifications
CTA w/1% Xylose | |
15mm x 103mm Tube | |
Agar | |
20 x 4 mL |