Learn More
Thermo Scientific™ HI Agar (Slant)
Cultivate and isolate fastidious and nonfastidious microorganisms Dehydrated Heart Infusion Agar.
$174.00 - $1426.00
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
R061012
|
Thermo Scientific™
R061012 |
20/Pk. |
Pack of 20 for $416.00
|
|
|||||
R061010
|
Thermo Scientific™
R061010 |
100/Pk. |
Case of 100 for $1,426.00
|
|
|||||
R01486
|
Thermo Scientific™
R01486 |
10/Pk. |
Pack of 10 for $174.00
|
|
|||||
Description
Cultivate and isolate fastidious and nonfastidious microorganisms using Thermo Scientific™ Remel™ Dehydrated Heart Infusion Agar. In the first media meat infusions were included for the cultivation of bacteria1. Huntoon prepared a medium using fresh beef heart and peptone which became known as Heart Infusion Agar. He demonstrated that without the addition of enrichment such as animal blood, it could be used to support the growth of nutritionally fastidious microorganisms.
- Ready to use - Solid media
- Used for mass cultivation of bacteria required in vaccine preparation and animal blood can also be added to enable determination of hemolytic reactions by organisms2,3
This medium contains beef heart infusion and casein peptone that supply nitrogenous compounds and amino acids necessary for the growth of nutritionally fastidious bacteria. Sodium chloride provides essential electrolytes and maintains osmotic equilibrium. Yeast extract is a source of energy and to enhance the bacterial growth it supplies B vitamins.
Specifications
Agar |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.