Learn More
Thermo Scientific™ Dermatophyte Test Medium (DTM)
Facilitate selective and differential isolation of pathogenic fungi (dermatophytes) from cutaneous sources using Dermatophyte Test Medium (DTM).
Supplier:  Thermo Scientific™ R10067
 View more versions of this product
Catalog No. R10067
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Facilitate selective and differential isolation of pathogenic fungi (dermatophytes) from cutaneous sources using Thermo Scientific™ Remel™ Dermatophyte Test Medium (DTM). This medium was formulated by Taplin et al. for use in locations where specialized training and microscopic examination is not available1. A pH indicator and three antimicrobial agents are added to the agar to provide a differential and selective medium for isolation of dermatophytes belonging to the genera Microsporum, Trichophyton, or Epidermophyton.
Contains a pH indicator.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dermatophyte Test Medium
1 oz. Bottle
Microbiology Media
10/Pk.