Learn More
Thermo Scientific™ Remel™ DNase Test Agar w/Methyl Green Available on GSA/VA Contract for Federal Government customers only.
Use for the detection of deoxyribonuclease activity in microorganisms including Staphylococci, enteric gram-negative bacilli and pseudomonads.
$163.07 - $299.78
Specifications
Description DNase Test Agar w/Methyl Green
View More Specs
Products 2
Catalog Number Mfr. No. Format Price Quantity & Availability  
Catalog Number Mfr. No. Format Price Quantity & Availability  
R01376 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R01376
12mm x 85mm Monoplate Pack of 10 for $163.07
Add to cart
R08342 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R08342
20mm x 113mm Tube Pack of 20 for $299.78
Add to cart
Description
Description
Thermo Scientific™ Remel DNase Test Agar with Methyl Green is for the detection of deoxyribonuclease activity in microorganisms including Staphylococci, enteric gram-negative bacilli and pseudomonads.
Methyl green dye is added to eliminate the need to flood the plate with HCl.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
DNase Test Agar w/Methyl Green