Learn More
Thermo Scientific™ Remel™ Easyplate Stacker
Thermo Scientific™ Remel Easyplate Stacker is for storing and carrying petri dishes.
Supplier: Thermo Scientific™ R55204
This item is not returnable.
View return policy
Description
Thermo Scientific™ Remel Easyplate Stacker is for storing and carrying petri dishes.
- Holds up to 56 plates
- Easy to use and carry
- Identification strip on side panel
- Helps prevent spills
- Color coded
Specifications
Easyplate Stacker | |
Dishes | |
Square |
Blue | |
56 | |
Yes |