Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Egg Yolk Tellurite 100% Available on GSA/VA Contract for Federal Government customers only.

Remel Egg Yolk Tellurite 100% is used with Baird Parker Agar Base.

$87.05 - $570.15

Specifications

Description Egg Yolk Tellurite 100%
View More Specs

Products 3
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
R450330 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R450330
Ea. 100mL Pack for $91.25
Add to cart
 
R450331 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R450331
10/Pk. 100mL Pack of 10 for $570.15
Add to cart
 
R450332
View Documents
Thermo Scientific™
R450332
10/Pk. 10mL Pack of 10 for $87.05
Add to cart
 
Description

Description

Remel Egg Yolk Tellurite 100% is used with Baird Parker Agar Base.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Egg Yolk Tellurite 100%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit