Learn More
Thermo Scientific™ Remel Esculin-PYR, mini-ID™ Enterococcus Tube Test Available on GSA/VA Contract for Federal Government customers only.
Superior to bacitracin sensitivity when confirming Group A Strep
Manufacturer:  Thermo Scientific™ R904205
 View more versions of this product
Catalog No. 23-246311
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Requires : Test requires use of PYR reagent (available separately)
Specifications Description & Specifications
Specifications
Non-waived
Contains materials for 50 Tests
30 to 240 min.
Streptococcus
50 Tests
Enterococcus Tube Test
Accessories