Thermo Scientific™ Remel Esculin-PYR, mini-ID™ Enterococcus Tube Test
Superior to bacitracin sensitivity when confirming Group A Strep
Manufacturer: Thermo Scientific™ R906401
 View more versions of this product
Catalog No. 23-246342
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Required Accessories : Test requires use of PYR reagent (available separately)
    Specifications
    Product Type Enterococcus Tube Test
    Quantity 50 Tests
    Test Time 30 to 240 min.
    Clia Complexity Non-waived
    Detectable Analytes Streptococcus
    Includes 15mL PYR reagent
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback