Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ Fluid D w/0.1% peptone, 0.1% Polysorbate 80
Thermo Scientific™ Remel Fluid D with 0.1% peptone, 0.1% Polysorbate 80 is used as diluting or rinsing fluid for sterility testing.
Supplier: Thermo Scientific™ R112303
This item is not returnable.
View return policy
Description
Thermo Scientific™ Remel Fluid D with 0.1% peptone, 0.1% Polysorbate 80 is used as diluting or rinsing fluid for sterility testing.
Specifications
Fluid D | |
United States | |
500 mL |
Borosilicate Bottle - Septum | |
Buffer Solution | |
10/Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Not intended for IVD use.
Spot an opportunity for improvement?
Provide Content Correction