Learn More
Thermo Scientific™ Remel™ Fraser Broth Base
Use with acriflavin, nalidixic acid, and ferric ammonium citrate for the selective enrichment of Listeria species.
$191.52 - $1002.75
Specifications
Description Fraser Broth Base
Format Powder
Packaging Ea.
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
R453482
Accessories View Documents
Thermo Scientific™
R453482
500g Each for $191.52
Add to cart
R453484
View Documents
Thermo Scientific™
R453484
2.5kg Each for $1,002.75
Add to cart
Description
Description
Remel Fraser Broth Base is for use with acriflavin, nalidixic acid, and ferric ammonium citrate for the selective enrichment of Listeria species.
Also available: Fraser Broth Additive, Part No. 450361.
Specifications
Specifications
Fraser Broth Base
Powder
Ea.
For Laboratory Use Only