Learn More
Thermo Scientific™ Remel™ GC Agar Base Available on GSA/VA Contract for Federal Government customers only.
Use with hemoglobin and GCHI enrichment for the isolation and cultivation of Neisseria/gonorrhoeae and other fastidious microorganisms.
$146.27 - $2150.00
Specifications
Description GC Agar Base
Format Powder
Packaging Ea.
View More Specs
Products 3
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
R453502 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R453502
Powder 500g Each for $146.27
Add to cart
R453504
View Documents
Thermo Scientific™
R453504
Powder 2.5Kg Each for $619.50
Add to cart
R453506
View Documents
Thermo Scientific™
R453506
Powder 10Kg Each for $2,150.00
Add to cart
Description
Description
Remel GC Agar Base is used with hemoglobin and GCHI enrichment for the isolation and cultivation of Neisseria/gonorrhoeae and other fastidious microorganisms.
Also available: GCHI Enrichment with Rehydrating Solution, Part No. R450411; VCN Selective Supplement, Part No. R450901; and VCNT Selective Supplement, Part No. R450921.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
GC Agar Base
Powder
Ea.
For Laboratory Use Only