Learn More
Thermo Scientific™ Remel™ Gram Crystal Violet Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Gram Crystal Violet is a primary stain in Gram stain procedures.
$49.56 - $216.83
Specifications
Certifications/Compliance CE Marked
View More Specs
Products 3
Catalog Number Mfr. No. Type Quantity Certifications/Compliance Price Quantity & Availability  
Catalog Number Mfr. No. Type Quantity Certifications/Compliance Price Quantity & Availability  
R40052 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R40052
250mL/Bottle Each CE Marked Each for $49.56
Add to cart
R40053 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R40053
250mL/Bottle 5/Pk. CE Marked Pack of 5 for $114.03
Add to cart
R40073 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R40073
1 gal. Each CE Marked Each for $216.83
Add to cart
Description
Description
Thermo Scientific™ Remel Gram Crystal Violet is a primary stain in Gram stain procedures.
  • For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
  • Selected Gram stains are CE marked
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
CE Marked