Learn More
Thermo Scientific™ Remel™ Gram Enhancer Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Gram Enhancer is used to reduce the red color of background debris to give better contrast to small gram-negative microorganisms.
$161.07 - $258.30
Products 2
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
R40232 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R40232
1 gal. Each
Each for $258.30
Add to cart
R40231 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R40231
5/pk. 250mL/Bottle
Pack for $161.07
Add to cart
Description
Description
Thermo Scientific™ Remel Gram Enhancer is used to reduce the red color of background debris to give better contrast to small gram-negative microorganisms.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.