Learn More
Thermo Scientific™ Remel™ Gram Safranin Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.
$55.65 - $206.75
Specifications
Certifications/Compliance CE Marked
View More Specs
Products 3
Catalog Number Mfr. No. Type Quantity Certifications/Compliance Price Quantity & Availability  
Catalog Number Mfr. No. Type Quantity Certifications/Compliance Price Quantity & Availability  
R40059 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R40059
250mL/Bottle 5/Pk. CE Marked
Pack of 5 for $118.23
Add to cart
R40079 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R40079
1 gal. Each CE Marked
Each for $206.75
Add to cart
R40058 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R40058
250mL/Bottle Each CE Marked
Each for $55.65
Add to cart
Description
Description
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.
  • For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
  • Selected Gram stains are CE marked
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
CE Marked