Learn More
Thermo Scientific™ Haemophilus Test Medium (HTM) Available on GSA/VA Contract for Federal Government customers only.
Perform effortless susceptibility testing of Haemophilus influenza with this ready-to-use medium which conforms to CLSI standards.
$89.04
Specifications
Description Haemophilus Test Medium
Packaging 10/Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Format Price Quantity & Availability  
Catalog Number Mfr. No. Format Price Quantity & Availability  
R01503 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R01503
15mm x 100mm Plate
Pack of 10 for $89.04
Add to cart
R04033 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R04033
15mm x 150mm Plate This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Perform effortless susceptibility testing of Haemophilus influenza with ready-to-use Thermo Scientific™ Remel™ Haemophilus Test Medium (HTM). The medium forms part of the recommended methods by the Clinical and Laboratory Standards Institute (CLSI)1. Haemophilus Test Medium has been specifically formulated for the susceptibility testing of Haemophilus influenzae. The results achieved using HTM have been found to be highly reproducible2,3. The transparency of the medium allows zones of inhibition to be easily read through the bottom of the plate.
The occurrence of resistant strains of Haemophilus influenza has increased in recent decades, underscoring the need for reliable susceptibility testing methods for this organisms. Modifications to standard susceptibility testing procedures have become necessary because of the fastidious nature of H. influenza. Complex growth-stimulating supplements, ordinarily used to grow Haemophilus spp., have been found to promote antagonisms of certain antibiotics and result in poor reproducibility4. Difficulties in interpreting inhibition zones may also arise.
Use Haemophilus Test Medium (HTM) for the susceptibility testing of Haemophilus influenzae. 
  • Conforms to CLSI standards
  • Produces highly reproducible results.
  • Transparent medium to facilitate the reading of zones of inhibition.
  • Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Haemophilus Test Medium
10/Pk.