Learn More
Thermo Scientific™ HI Broth (5mL)
Cultivate and isolate fastidious and nonfastidious microorganisms using Heart Infusion Broth.
Supplier: Thermo Scientific™ R061030
Description
Cultivate and isolate fastidious and nonfastidious microorganisms using Thermo Scientific™ Remel™ Heart Infusion Broth. In the first media meat infusions were included for the cultivation of bacteria1. Huntoon prepared a medium using fresh beef heart and peptone which was known as Heart Infusion Broth. He demonstrated that without the addition of enrichment such as animal blood, it could be used to support the growth of nutritionally fastidious microorganisms.
- Used for mass cultivation of bacteria required in vaccine preparation and animal blood can also be added to enable determination of hemolytic reactions by organisms2,3.
- Ready to use convenience of prepared media.
This medium contains beef heart infusion and casein peptone that supply nitrogenous compounds and amino acids necessary for the growth of nutritionally fastidious bacteria. Sodium chloride provides essential electrolytes and maintains osmotic equilibrium. Yeast extract is a source of energy and to enhance the bacterial growth it supplies B vitamins.
Specifications
Industrial Reference: BAM, COMPF. | |
15mm x 103mm Tube | |
100 x 5 mL |
Heart Infusion Broth | |
Microbiology Media |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.