Learn More
Thermo Scientific™ KBE (Kanamycin Bile Esculin) Agar
Selectively isolate and presumptively identify Bacteroides fragilis group using KBE Agar (Kanamycin Bile Esculin Agar).
Supplier:  Thermo Scientific™ R01511
 View more versions of this product
Catalog No. R01511
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
This item is not returnable. View return policy
Description
Description
Selectively isolate and presumptively identify Bacteroides fragilis group using Thermo Scientific™ Remel™ KBE Agar (Kanamycin Bile Esculin Agar). Chan and Porschen formulated KBE Agar to selectively isolate and presumptively identify the B. fragilis group based on resistance to bile and kanamycin and hydrolysis of esculin1,2. Clinical specimens for anaerobic culture frequently contain mixed flora, including facultative gram-negative rods that readily overgrow the anaerobes, so selective media are recommended for isolation of anaerobes3.
Members of the Bacteroides fragilis group are the anaerobic bacteria most frequently isolated in human clinical infections4. This group includes B. fragilis, B. thetaiotaomicron, B. ovatus, B. distasonis, B. vulgatus, and others. These members of the B. fragilis group hydrolyze esculin to produce esculetin.
Selectively isolate and presumptively identify members of the B. fragilis group using KBE Agar.
  • Ready to use – Convenience of prepared media
  • Easy to interpret - Brown-black complex in the agar surrounding the colony
  • Selective media - Addition of kanamycin and bile makes this medium selective
    • Kanamycin (1000μg/ml) inhibits facultative anaerobes and anaerobic gram-negative bacteria other than B. fragilis group
    • Bile (20%) stimulates B. fragilis but inhibits other anaerobic bacteria
Ferric ammonium citrate present in the medium reacts with esculetin to produce a brown-black complex in the agar surrounding the colony. Addition of kanamycin and bile to this medium makes it selective. Vitamin K and hemin are growth factors. Hemin also allows testing for catalase production5.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KBE Agar
10mm x 100mm Spacesaver Plate
Agar
15/Pk.