Learn More
Thermo Scientific™ Remel™ LB Agar (Luria-Bertani)
Thermo Scientific™ Remel LB Agar (Luria-Bertani) is for the cultivation and maintenance of Escherichia coli for genetic and molecular studies.
$83.06
Specifications
Description LB Agar
Packaging 10/Pk.
View More Specs
Products 1
Catalog Number Mfr. No. Format Price Quantity & Availability  
Catalog Number Mfr. No. Format Price Quantity & Availability  
R110203
View Documents
Thermo Scientific™
R110203
12mm x 85mm Monoplate Pack of 10 for $83.06
Add to cart
Description
Description
Thermo Scientific™ Remel LB Agar (Luria-Bertani) is for the cultivation and maintenance of Escherichia coli for genetic and molecular studies.
Specifications
Specifications
LB Agar
10/Pk.