Learn More
Thermo Scientific™ Remel™ Lowenstein Jensen Base
Use with fresh eggs and glycerol for isolation and differentiation of Mycobacterium species.
$1183.00
Specifications
Description Lowenstein Jensen Base
View More Specs
Products 1
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
R453754
View Documents
Thermo Scientific™
R453754
Lowenstein Jensen Base 2.5kg
Each for $1,183.00
Add to cart
Description
Description
The Thermo Scientific™ Remel Lowenstein Jensen Base is used with fresh eggs and glycerol for the isolation and differentiation of Mycobacterium species.
Specifications
Specifications
Lowenstein Jensen Base
SDS
For Laboratory Use Only