Learn More
Thermo Scientific™ Remel™ Lowenstein-Jensen Gruft (LJ) w/penicillin, nalidixic acid Available on GSA/VA Contract for Federal Government customers only.
The Thermo Scientific™ Remel Lowenstein-Jensen Gruft (LJ) with penicillin, nalidixic acid for the selective isolation of Mycobacterium spp.
$231.53 - $2897.27
Specifications
Description Lowenstein-Jensen Gruft. (LJ) w/penicillin, nalidixic acid
View More Specs
Products 4
Catalog Number Mfr. No. Format Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Format Packaging Price Quantity & Availability  
R08520 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R08520
20mm x 113mm Tube - Slant 100/Pk.
Case of 100 for $911.40
Add to cart
R08522 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R08522
20mm x 113mm Tube - Slant 20/Pk.
Pack of 20 for $231.53
Add to cart
R10102
View Documents
Thermo Scientific™
R10102
1 oz. Bottle 100/Pk.
Pack of 100 for $2,897.27
Add to cart
R10103
View Documents
Thermo Scientific™
R10103
1 oz. Bottle 10/Pk.
Pack of 10 for $263.51
Add to cart
Description
Description
The Thermo Scientific™ Remel Lowenstein-Jensen Gruft (LJ) with penicillin, nalidixic acid is for the selective isolation of Mycobacterium spp.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Lowenstein-Jensen Gruft. (LJ) w/penicillin, nalidixic acid