Learn More
Thermo Scientific™ Remel™ m-ENDO Agar LES Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel m-ENDO Agar LES is used for cultivation and enumeration of coliform bacteria from water by membrane filtration method.
$139.65 - $146.90
Specifications
Description m-ENDO Agar LES
Packaging 10/Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Format Price Quantity & Availability  
Catalog Number Mfr. No. Format Price Quantity & Availability  
R01425
View Documents
Thermo Scientific™
R01425
15mm x 60mm Plate Pack of 10 for $139.65
Add to cart
R01426 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R01426
12mm x 85mm Monoplate Pack of 10 for $146.90
Add to cart
Description
Description
Thermo Scientific™ Remel m-ENDO Agar LES is used for cultivation and enumeration of coliform bacteria from water by membrane filtration method.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
m-ENDO Agar LES
10/Pk.