Learn More
Thermo Scientific™ Malt Extract Agar
Isolate and enumerate yeast and molds from dairy products and other foodstuffs using Malt Extract Agar.
Supplier: Thermo Scientific™ R01567
Description
Isolate and enumerate yeast and molds using Thermo Scientific™ Remel™ Malt Extract Agar. The utilization of malt or malt extracts for the propagation of yeast and molds is quite common. Reddish described the use of malt extract as a substitute for beer wort in a culture medium1. Thom and Church developed Malt Extract Agar, a modification of Reddish’s formula, as a general cultivation medium for Aspergilli2. Malt Extract Agar is recommended for the detection of yeasts and molds in dairy products and other foodstuffs3-5.
Isolate and enumerate yeast and molds in dairy products and other foodstuffs using Malt Extract Agar. Both fresh and processed products are subject to fungal spoilage and even heat-processed foods may be affected. Certain food-borne molds and yeasts may also elicit allergic reactions or may cause infections.
This medium contains high concentration of maltose and this high carbohydrate content ensures rapid growth of yeasts and molds. Dextrin and glycerol are carbon sources and gelatin peptone is a source of nitrogen. The acidic pH of this medium is optimum for the growth of yeasts and molds while inhibiting the growth of bacteria. Selectivity can be increased by further lowering the pH with the addition, after sterilization, of lactic acid.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Industrial reference: COMPF. | |
12mm x 85mm Monoplate | |
10/Pk. |
Malt Extract Agar | |
Agar |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Not intended for IVD use.