Learn More
Thermo Scientific™ Remel™ Micro-ID™ Listeria Identification System
Self-contained unit with reagent impregnated paper disks for fifteen biochemical tests for differentiation of Listeria in 24 hours.
Manufacturer:  Thermo Scientific™ R38370
Catalog No. R38370
Add to cart
Description
Description
Remel Micro-ID Listeria Identification System is a self-contained unit with reagent impregnated paper disks for fifteen biochemical tests for differentiation of Listeria in 24 hours.
Specifications
Specifications
Micro-ID™ Listeria Identification System
10 Units/Pk.
Documents
Accessories