Learn More
Thermo Scientific™ Remel™ Microring™ XV System
GSA_VA
Thermo Scientific™ Remel Microring XV is an identification system for Haemophilus spp. using X, V and XV factors plus porphyrin test.
Manufacturer:  Thermo Scientific™ R64502
Catalog No. R64502
Add to cart
This item is not returnable. View return policy
Description
Description
Thermo Scientific™ Remel Microring XV is an identification system for Haemophilus spp. using X, V and XV factors plus porphyrin test.
  • Reagent impregnated filter paper ring
  • Medical Wire and Equipment
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Microring™ XV
Haemophilus
Microring XV
50 Rings/Pk.