Learn More
Thermo Scientific™ Remel™ Middlebrook 7H9 Broth w/Polysorbate 80 (Tellurite Reduction Medium) Available on GSA/VA Contract for Federal Government customers only.
Use for the differentiation of Mycobacterium spp. based on tellurite reduction.
$267.75 - $1392.30
Specifications
Description Middlebrook 7H9 Broth with Polysorbate 80 Tellurite Reduction Medium
Format 20mm x 150mm Tube
View More Specs
Products 2
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
R09556 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R09556
100/Pk. Case of 100 for $1,392.30
Add to cart
R09558 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R09558
20/Pk. Pack of 20 for $267.75
Add to cart
Description
Description
Thermo Scientific™ Remel Middlebrook 7H9 Broth with Polysorbate 80 (Tellurite Reduction Medium) is for the differentiation of Mycobacterium spp. based on tellurite reduction.
Requires:
Requires Potassium Tellurite Reagent, REF R21254.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Middlebrook 7H9 Broth with Polysorbate 80 Tellurite Reduction Medium
20mm x 150mm Tube