Learn More
Thermo Scientific™ Remel mini-ID™ Fastidious Fermenter Rapid Tube Tests Available on GSA/VA Contract for Federal Government customers only.
Rapid Carbohydrate Degradation (RCD) microtube test
Manufacturer:  Thermo Scientific™ R907302
 View more versions of this product
Catalog No. 23-246325
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
N. sicca, N. subflava, N. mucosa and N. flavescens
50 Tests
4 to 24 hr.
Sucrose/Fructose/Nitrate-Esculin MN2 Kit