Learn More
Thermo Scientific™ Remel™ Motility GI Medium Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Motility GI Medium is for the motility testing of Enterobacteriaceae.
$247.80 - $1686.30
Specifications
Description Motility GI Medium
Format 15mm x 103mm Tube
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
R061400 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R061400
20 x 5mL Pack of 20 for $247.80
Add to cart
R061398
View Documents
Thermo Scientific™
R061398
100 x 5mL Case of 100 for $1,686.30
Add to cart
Description
Description
Thermo Scientific™ Remel Motility GI Medium is for the motility testing of Enterobacteriaceae.
  • Semi-solid gelatin heart infusion medium
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Motility GI Medium
15mm x 103mm Tube