Learn More
Thermo Scientific™ MH Broth w/Cations
Cultivate a wide variety of microorganisms using Mueller Hinton Broth with Cations.
Supplier: Thermo Scientific™ R112475
Description
Cultivate a wide variety of microorganisms using Thermo Scientific™ Remel™ Mueller Hinton Broth with Cations. Mueller Hinton formulated this medium as a protein-free medium for the purpose of isolating pathogenic Neisseria1. This was later used for testing gonococci and other organisms for susceptibility to sulfonamides2,3. More recently, MH Broth with Cations has become useful as a general-purpose cultivation broth.
Cultivate a wide variety of microorganisms with Mueller Hinton Broth with Cations.
- Divalent cations calcium and magnesium enrich the medium and serve to support the growth of fastidious microorganisms.
- Ready-to-use – Convenience of prepared media.
Mueller Hinton Broth is prepared with beef extract and acid digest of casein that supply amino acids, nitrogenous substances, and other nutrients which are necessary for bacterial growth. Starch serves as a growth factor and a protective colloid that neutralizes toxic products which may be formed during the growth of bacteria. Calcium and Magnesium are divalent cations that enrich the medium.
Specifications
Mueller Hinton Broth w/Cations (Ca, Mg) | |
Microbiology Media | |
4 x 500 mL |
Borosilicate Bottle - 500mL | |
4/Pk. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.