Learn More
Thermo Scientific™ MH Broth
Cultivate a wide variety of microorganisms using Mueller Hinton Broth.
Supplier: Thermo Scientific™ R08594
Description
Cultivate a wide variety of microorganisms using Thermo Scientific™ Remel™ Mueller Hinton Broth. Mueller Hinton formulated this medium as a protein-free medium for the purpose of isolating pathogenic Neisseria1. MH Broth was later used for testing gonococci and other organisms for susceptibility to sulfonamides2,3.
Cultivate a wide variety of microorganisms using Mueller Hinton Broth.
- Recommended by FDA, World Health Organization (WHO) and NCCLS for testing most commonly encountered aerobic and facultative anaerobic bacteria in food and clinical material4,5.
- Ready-to-use – Convenience of prepared media.
Mueller Hinton Broth is prepared with beef extract and acid digest of casein that supply amino acids, nitrogenous substances, vitamins, carbon, sulphur and other nutrients which are necessary for bacterial growth. Starch serves as a growth factor and a protective colloid that neutralizes toxic products which may be formed during the growth of bacteria.
Specifications
Mueller Hinton Broth | |
Microbiology Media | |
100 x 10 mL |
20mm x 113mm Tube | |
100/Pk. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.