Learn More
Thermo Scientific™ MH Broth
Cultivate a wide variety of microorganisms using Mueller Hinton Broth.
Supplier: Thermo Scientific™ R112474
Description
Cultivate a wide variety of microorganisms using Thermo Scientific™ Remel™ Mueller Hinton Broth. Mueller Hinton formulated this medium as a protein-free medium for the purpose of isolating pathogenic Neisseria1. MH Broth was later used for testing gonococci and other organisms for susceptibility to sulfonamides2,3.
Cultivate a wide variety of microorganisms using Mueller Hinton Broth.
- Recommended by FDA, World Health Organization (WHO) and NCCLS for testing most commonly encountered aerobic and facultative anaerobic bacteria in food and clinical materi
Specifications
Mueller Hinton Broth | |
Microbiology Media | |
4 x 500 mL |
Borosilicate Bottle | |
4/Pk. |