Learn More
Thermo Scientific™ Remel™ NAC Attack
Catalog No. p-4522359
Click to view available options
Quantity:
5 x 50 mL
5 x 100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Remel NAC Attack is for digestion and decontamination procedures for AFB cultures.
Remel NAC Attack is for digestion and decontamination procedures for AFB cultures.
Includes:
Five sealed ampules of lyophilized N-acetyl-L-cysteine; five bottles TB Base Digestant.
TRUSTED_SUSTAINABILITY
Specifications
Description NAC Attack
Product Type Bacterial and Fungal Isolation
Quantity 5 x 50 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.