Learn More
Thermo Scientific™ Remel™ Nitrate Broth Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Nitrate Broth is for the determination of nitrate reduction by bacteria.
$199.71 - $947.10
Specifications
Certifications/Compliance Industrial Reference: BAM, COMPF, SMD.
Description Nitrate Broth
View More Specs
Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
R061532 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R061532
20 x 5mL Pack of 20 for $199.71
Add to cart
R061536 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R061536
20 x 7mL Pack of 20 for $199.82
Add to cart
R061530
View Documents
Thermo Scientific™
R061530
100 x 5mL Case of 100 for $947.10
Add to cart
R061534 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R061534
100 x 7mL Pack of 100 for $858.90
Add to cart
Description
Description
Thermo Scientific™ Remel Nitrate Broth is for the determination of nitrate reduction by bacteria.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Industrial Reference: BAM, COMPF, SMD.
Nitrate Broth