Learn More
Thermo Scientific™ Remel™ OF King Medium
Thermo Scientific™ Remel OF King Medium is for the determination of oxidative and fermentative metabolism of carbohydrates by gram-negative bacilli.
Manufacturer:  Thermo Scientific™ R061919
 View more versions of this product
Catalog No. R061919
Add to cart
Description
Description
Thermo Scientific™ Remel OF King Medium is for the determination of oxidative and fermentative metabolism of carbohydrates by gram-negative bacilli.
Specifications
Specifications
OF King Medium w/1% Dextrose; Format
20 x 5mL
20/Pk.
Media
15mm x 103mm Tube