Learn More
Thermo Scientific™ Remel™ Phadebact™ Haemophilus Test
A slide co-agglutination test used for confirmatory identification of Haemophilus influenzae.
Manufacturer:  Thermo Scientific™ R60901
Catalog No. R60901
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
This item is not returnable. View return policy
Description
Description
Phadebact™ Haemophilus Test is a slide co-agglutination test used for confirmatory identification of Haemophilus influenzae.
Includes:
1 reagent for Type b; 1 polyvalent reagent for Types a, c, d, e and f; droppers; disposable slides
Phadebact™ is a registered trademark of MKL Diagnostics AB.
Specifications
Specifications
Phadebact Haemophilus Test
Haemophilis Testing Kit
50 Tests/Kit
Haemophilus influenzae
1 reagent for Type b; 1 polyvalent reagent for Types a, c, d, e, and f; droppers; disposable slides