Learn More
Thermo Scientific™ Remel™ Phadebact™ Monoclonal GC Test Available on GSA/VA Contract for Federal Government customers only.
A slide co-agglutination test used for confirmatory identification of Neisseria gonorrhoeae.
Manufacturer:  Thermo Scientific™ R60802
Catalog No. R60802
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Phadebact™ Monoclonal GC Test is a slide co-agglutination test used for confirmatory identification of Neisseria gonorrhoeae.
Includes:
1 WI reagent; 1 WII/WIII reagent; droppers; disposable slides
Phadebact™ is a registered trademark of MKL Diagnostics AB.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Healthcare
Phadebact Monoclonal GC Test
1 WI reagent; 1 WII/WIII reagent; droppers; disposable slides
50 Tests/Kit