Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Phenylalanine Agar Available on GSA/VA Contract for Federal Government customers only.

Thermo Scientific™ Remel Phenylalanine Agar is for differentiation of enteric bacilli based on deamination of phenylalanine.

$188.16 - $788.03

Specifications

Certifications/Compliance Industrial Reference: BAM.
Description Phenylalanine Agar
Format 15mm x 103mm Tube - Slant
View More Specs

Products 2
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
R062180 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R062180
100/Pk.
Case of 100 for $788.03
Add to cart
 
R062182 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R062182
20/Pk.
Pack of 20 for $188.16
Add to cart
 
Description

Description

Thermo Scientific™ Remel Phenylalanine Agar is for differentiation of enteric bacilli based on deamination of phenylalanine.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Industrial Reference: BAM.
Phenylalanine Agar
15mm x 103mm Tube - Slant
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit